Assay Details
Target Gene Details
Entrez Gene ID: | 153478 |
Gene Name: | pleckstrin homology and RhoGEF domain containing G4B |
Gene Aliases: |
- |
Location: |
Chr.5:54065-189972 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLEKHG4B | XM_011513971.2 | XP_011512273.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv822925 | Chr.5:49928 - 185592 on Build GRCh38 | Gain |
|
| esv2752083 | Chr.5:28951 - 346521 on Build GRCh38 | Gain |
|
| dgv5519n100 | Chr.5:71184 - 138345 on Build GRCh38 | Gain |
|
| dgv5514n100 | Chr.5:15520 - 138345 on Build GRCh38 | Gain |
|
| nsv10643 | Chr.5:59149 - 119560 on Build GRCh38 | Gain+Loss |
|
| nsv509040 | Chr.5:59249 - 86835 on Build GRCh38 | Insertion |
|
| dgv5518n100 | Chr.5:63270 - 115984 on Build GRCh38 | Gain |
|
| nsv1031835 | Chr.5:46687 - 138345 on Build GRCh38 | Gain |
|
| esv3603720 | Chr.5:40419 - 176322 on Build GRCh38 | Loss |
|
| esv2759315 | Chr.5:17263 - 595840 on Build GRCh38 | Loss |
|
| nsv461885 | Chr.5:27565 - 118291 on Build GRCh38 | Gain |
|
| dgv5517n100 | Chr.5:19572 - 164078 on Build GRCh38 | Loss |
|
| dgv9471n54 | Chr.5:12225 - 141544 on Build GRCh38 | Loss |
|
| dgv5516n100 | Chr.5:17630 - 81382 on Build GRCh38 | Loss |
|
| esv26253 | Chr.5:73864 - 76275 on Build GRCh38 | Loss |
|
| nsv950616 | Chr.5:71286 - 94685 on Build GRCh38 | Deletion |
|
| nsv441935 | Chr.5:68560 - 115996 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73730595] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map