Assay Details
Target Gene Details
Entrez Gene ID: | 101929696 |
Gene Name: | uncharacterized LOC101929696 |
Gene Aliases: |
- |
Location: |
Chr.5:139741347-139746223 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929696 | NR_105052.2 | |||
| NR_105053.2 | ||||
| BX105638.1 | ||||
| HY044566.1 |
Target Gene Details
Entrez Gene ID: | 84249 |
Gene Name: | pleckstrin and Sec7 domain containing 2 |
Gene Aliases: |
EFA6C |
Location: |
Chr.5:139683980-139844466 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSD2 | XM_011537696.2 | XP_011535998.1 | ||
| XM_017009975.1 | XP_016865464.1 | |||
| XM_017009976.1 | XP_016865465.1 | |||
| XM_017009977.1 | XP_016865466.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv818386 | Chr.5:139708492 - 139765499 on Build GRCh38 | Gain |
|
| nsv519808 | Chr.5:139396890 - 139805924 on Build GRCh38 | Loss |
|
| nsv462460 | Chr.5:139553566 - 139765464 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map