Assay Details
Target Gene Details
Entrez Gene ID: | 90249 |
Gene Name: | unc-5 netrin receptor A |
Gene Aliases: |
UNC5H1 |
Location: |
Chr.5:176810559-176880898 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UNC5A | NM_133369.2 | NP_588610.2 | ||
| XM_006714927.1 | XP_006714990.1 | |||
| XM_006714928.1 | XP_006714991.1 | |||
| XM_011534686.1 | XP_011532988.1 | |||
| XM_011534687.2 | XP_011532989.1 | |||
| AK131380.1 | BAD18531.1 | |||
| BC157824.1 | ||||
| BI818609.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2763524 | Chr.5:176729717 - 176857712 on Build GRCh38 | Gain |
|
| nsv950724 | Chr.5:176766400 - 176890599 on Build GRCh38 | Deletion |
|
| nsv509100 | Chr.5:176770480 - 176835979 on Build GRCh38 | Insertion |
|
| dgv10181n54 | Chr.5:176808074 - 176883197 on Build GRCh38 | Loss |
|
| esv3607674 | Chr.5:176753572 - 176873255 on Build GRCh38 | Gain |
|
| nsv462557 | Chr.5:176741506 - 176866702 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map