Assay Details
Target Gene Details
Entrez Gene ID: | 3274 |
Gene Name: | histamine receptor H2 |
Gene Aliases: |
H2R |
Location: |
Chr.5:175657762-175710756 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HRH2 | NM_001131055.1 | NP_001124527.1 | ||
| XM_005265904.3 | XP_005265961.1 | |||
| XM_006714864.3 | XP_006714927.1 | |||
| XM_006714865.3 | XP_006714928.1 | |||
| XM_011534548.2 | XP_011532850.1 | |||
| XM_011534549.2 | XP_011532851.1 | |||
| XM_017009426.1 | 1 | 146 | XP_016864915.1 | |
| BC054510.2 | AAH54510.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469707 | Chr.5:175516474 - 175693143 on Build GRCh38 | Loss |
|
| nsv600335 | Chr.5:175653226 - 175714243 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73337491,rs73802987] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map