Assay Details
Target Gene Details
Entrez Gene ID: | 3796 |
Gene Name: | kinesin family member 2A |
Gene Aliases: |
CDCBM3, HK2, KIF2 |
Location: |
Chr.5:62306162-62387184 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIF2A | NM_001098511.2 | NP_001091981.1 | ||
| NM_001243952.1 | NP_001230881.1 | |||
| NM_001243953.1 | NP_001230882.1 | |||
| NM_004520.4 | NP_004511.2 | |||
| AK297347.1 | ||||
| AK307880.1 | ||||
| AK315866.1 | ||||
| DB452624.1 | ||||
| DB456309.1 | ||||
| EF560716.1 | ||||
| EF560728.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3570170 | Chr.5:62277474 - 62311441 on Build GRCh38 | Loss |
|
| dgv5689n100 | Chr.5:62155965 - 62735538 on Build GRCh38 | Gain |
|
| nsv830318 | Chr.5:62119886 - 62327493 on Build GRCh38 | Gain |
|
| esv2759345 | Chr.5:62107311 - 62453085 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs111244915] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map