Assay Details
Target Gene Details
Entrez Gene ID: | 728264 |
Gene Name: | cardiac mesoderm enhancer-associated non-coding RNA |
Gene Aliases: |
CARMEN, MIR143HG |
Location: |
Chr.5:149406845-149432836 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CARMN | NR_105059.1 | 5 | 787 | |
| NR_105060.1 | 4 | 723 | ||
| HQ848934.1 | 5 | 787 | ||
| HQ848935.1 | 4 | 723 |
Target Gene Details
Entrez Gene ID: | 406935 |
Gene Name: | microRNA 143 |
Gene Aliases: |
MIRN143, mir-143 |
Location: |
Chr.5:149428918-149429023 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR143 | NR_029684.1 | |||
| AJ535834.1 |

Genomic Map