Assay Details
Target Gene Details
Entrez Gene ID: | 3459 |
Gene Name: | interferon gamma receptor 1 |
Gene Aliases: |
CD119, IFNGR, IMD27A, IMD27B |
Location: |
Chr.6:137197484-137220351 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IFNGR1 | NM_000416.2 | NP_000407.1 | ||
| XM_006715470.2 | XP_006715533.1 | |||
| XM_006715471.2 | XP_006715534.1 | |||
| XM_011535793.2 | XP_011534095.1 | |||
| XM_011535794.1 | XP_011534096.1 | |||
| XM_017010827.1 | XP_016866316.1 | |||
| AF056979.1 | AAK30623.1 | |||
| AK222803.1 | BAD96523.1 | |||
| AK297052.1 | ||||
| AK312674.1 | ||||
| BC005333.1 | AAH05333.1 | |||
| BT006814.1 | AAP35460.1 | |||
| EF535103.1 | ||||
| J03143.1 | AAA52731.1 | |||
| KU178144.1 | ||||
| KU178145.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759473 | Chr.6:137138391 - 137252201 on Build GRCh38 | Loss |
|
| nsv470863 | Chr.6:137080640 - 138014159 on Build GRCh38 | Loss |
|
| esv32557 | Chr.6:137186345 - 137241212 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map