Assay Details
Target Gene Details
Entrez Gene ID: | 1026 |
Gene Name: | cyclin dependent kinase inhibitor 1A |
Gene Aliases: |
CAP20, CDKN1, CIP1, MDA-6, P21, SDI1, WAF1, p21CIP1 |
Location: |
Chr.6:36676460-36687339 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDKN1A | NM_001220777.1 | NP_001207706.1 | ||
| NM_001291549.1 | NP_001278478.1 | |||
| NM_078467.2 | NP_510867.1 | |||
| AK298901.1 | ||||
| BU154329.1 | ||||
| BU193165.1 | ||||
| DA702185.1 | ||||
| DA838987.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1015689 | Chr.6:36550916 - 39052434 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map