Assay Details
Target Gene Details
Entrez Gene ID: | 441172 |
Gene Name: | uncharacterized LOC441172 |
Gene Aliases: |
- |
Location: |
Chr.6:138691668-138697288 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FLJ46906 | NR_033896.1 | |||
| AK128874.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2752090 | Chr.6:138395100 - 138705640 on Build GRCh38 | Gain |
|
| nsv1122139 | Chr.6:138692463 - 138693563 on Build GRCh38 | Deletion |
|

Genomic Map