Assay Details
Target Gene Details
Entrez Gene ID: | 8418 |
Gene Name: | cytidine monophospho-N-acetylneuraminic acid hydroxylase, pseudogene |
Gene Aliases: |
CMAH, CSAH |
Location: |
Chr.6:25081067-25138392 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CMAHP | NR_027626.1 | |||
| AK303297.1 | ||||
| DA976765.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470806 | Chr.6:25082034 - 25155969 on Build GRCh38 | Loss |
|
| nsv1025694 | Chr.6:25136632 - 25309222 on Build GRCh38 | Gain |
|
| nsv830611 | Chr.6:25039146 - 25198884 on Build GRCh38 | Loss |
|
| nsv601166 | Chr.6:25084490 - 25165258 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75800218] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map