Assay Details
Target Gene Details
Entrez Gene ID: | 2037 |
Gene Name: | erythrocyte membrane protein band 4.1 like 2 |
Gene Aliases: |
4.1-G, 4.1G |
Location: |
Chr.6:130829111-131063322 on Build GRCh38 |
Assay Gene Location: | Within Intron 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPB41L2 | XM_017010353.1 | XP_016865842.1 |
Target Gene Details
Entrez Gene ID: | 100507203 |
Gene Name: | small leucine rich protein 1 |
Gene Aliases: |
- |
Location: |
Chr.6:130827405-130837135 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMLR1 | NM_001195597.1 | NP_001182526.1 | ||
| AK091900.1 | ||||
| BX095531.1 |
More Information
Set Membership: |
|

Genomic Map