Assay Details
Target Gene Details
Entrez Gene ID: | 441172 |
Gene Name: | uncharacterized LOC441172 |
Gene Aliases: |
- |
Location: |
Chr.6:138691668-138697288 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FLJ46906 | NR_033896.1 |
Target Gene Details
Entrez Gene ID: | 57224 |
Gene Name: | NHS like 1 |
Gene Aliases: |
C6orf63 |
Location: |
Chr.6:138422043-138692616 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NHSL1 | XM_011535976.1 | XP_011534278.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2752090 | Chr.6:138395100 - 138705640 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
