Assay Details
Target Gene Details
Entrez Gene ID: | 83699 |
Gene Name: | SH3 domain binding glutamate rich protein like 2 |
Gene Aliases: |
- |
Location: |
Chr.6:79537561-79703670 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SH3BGRL2 | XM_011536282.2 | XP_011534584.1 | ||
| XM_017011344.1 | XP_016866833.1 | |||
| XM_017011345.1 | XP_016866834.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv463906 | Chr.6:79531234 - 79603580 on Build GRCh38 | Loss |
|
| esv3890891 | Chr.6:79406146 - 79635231 on Build GRCh38 | Gain |
|
| esv3576196 | Chr.6:79528311 - 79781361 on Build GRCh38 | Gain |
|
| nsv830707 | Chr.6:79424908 - 79615454 on Build GRCh38 | Gain |
|
| nsv1027388 | Chr.6:79403732 - 79646256 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map