Assay Details
Target Gene Details
Entrez Gene ID: | 221756 |
Gene Name: | serpin family B member 9 pseudogene 1 |
Gene Aliases: |
SERPINB9P |
Location: |
Chr.6:2854657-2876510 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPINB9P1 | NR_033851.1 | |||
| BC025340.1 | AAH25340.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830575 | Chr.6:2847376 - 3035682 on Build GRCh38 | Loss |
|

Genomic Map