Assay Details
Target Gene Details
Entrez Gene ID: | 100113407 |
Gene Name: | transmembrane protein 170B |
Gene Aliases: |
- |
Location: |
Chr.6:11538042-11583524 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM170B | NM_001100829.2 | NP_001094299.1 | ||
| DB092520.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv600989 | Chr.6:11509090 - 11581757 on Build GRCh38 | Loss |
|

Genomic Map