Assay Details
Target Gene Details
Entrez Gene ID: | 55122 |
Gene Name: | akirin 2 |
Gene Aliases: |
C6orf166, FBI1, dJ486L4.2 |
Location: |
Chr.6:87674860-87702267 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AKIRIN2 | NM_018064.3 | NP_060534.1 | ||
| AK001204.1 | BAA91551.1 | |||
| AK222701.1 | BAD96421.1 | |||
| BC000764.2 | AAH00764.1 | |||
| BC003042.1 | AAH03042.1 | |||
| BC005051.1 | AAH05051.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3571280 | Chr.6:87691615 - 87699688 on Build GRCh38 | Loss |
|
| dgv6085n100 | Chr.6:87443361 - 87700332 on Build GRCh38 | Gain |
|
| nsv950478 | Chr.6:87683683 - 87703282 on Build GRCh38 | Duplication |
|
| esv2759454 | Chr.6:87649270 - 87731755 on Build GRCh38 | Loss |
|
| nsv7941 | Chr.6:87693837 - 87703877 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map