Assay Details
Target Gene Details
Entrez Gene ID: | 84498 |
Gene Name: | family with sequence similarity 120B |
Gene Aliases: |
CCPG, KIAA1838, PGCC1, dJ894D12.1 |
Location: |
Chr.6:170290703-170407748 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM120B | NM_001286379.1 | NP_001273308.1 | ||
| NM_001286380.1 | NP_001273309.1 | |||
| NM_001286381.1 | NP_001273310.1 | |||
| NM_032448.2 | NP_115824.1 | |||
| XM_011536185.2 | XP_011534487.1 | |||
| XM_017011358.1 | XP_016866847.1 | |||
| XM_017011359.1 | XP_016866848.1 | |||
| XM_017011360.1 | XP_016866849.1 | |||
| AB058741.1 | ||||
| AK056435.1 | ||||
| AK294415.1 | ||||
| AK296730.1 | ||||
| AK296825.1 | ||||
| AK299890.1 | ||||
| AL832883.1 | ||||
| BC012177.1 | AAH12177.1 | |||
| DQ873695.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759497 | Chr.6:170272663 - 170700831 on Build GRCh38 | Gain+Loss |
|
| esv3611733 | Chr.6:170307545 - 170312411 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map