Assay Details
Target Gene Details
Entrez Gene ID: | 84498 |
Gene Name: | family with sequence similarity 120B |
Gene Aliases: |
CCPG, KIAA1838, PGCC1, dJ894D12.1 |
Location: |
Chr.6:170290703-170407748 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM120B | NM_001286379.1 | NP_001273308.1 | ||
| NM_001286380.1 | NP_001273309.1 | |||
| AK294415.1 | ||||
| AK299890.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759497 | Chr.6:170272663 - 170700831 on Build GRCh38 | Gain+Loss |
|
| nsv605554 | Chr.6:170263653 - 170308522 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map