Assay Details
Target Gene Details
Entrez Gene ID: | 728464 |
Gene Name: | methyltransferase like 24 |
Gene Aliases: |
C6orf186, dJ71D21.2 |
Location: |
Chr.6:110245946-110358272 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| METTL24 | NM_001123364.1 | NP_001116836.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs79612029] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map