Assay Details
Target Gene Details
Entrez Gene ID: | 80129 |
Gene Name: | coiled-coil domain containing 170 |
Gene Aliases: |
C6orf97, bA282P11.1 |
Location: |
Chr.6:151494040-151621193 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC170 | NM_025059.3 | NP_079335.2 | ||
| AK026958.1 | BAB15602.1 | |||
| AK225330.1 | ||||
| BC060803.1 | AAH60803.1 | |||
| BI562667.1 | ||||
| DB232703.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1016522 | Chr.6:151412750 - 151534573 on Build GRCh38 | Gain |
|
| esv3576236 | Chr.6:151411765 - 151532455 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79112354] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map