Assay Details
Target Gene Details
Entrez Gene ID: | 221756 |
Gene Name: | serpin family B member 9 pseudogene 1 |
Gene Aliases: |
SERPINB9P |
Location: |
Chr.6:2854657-2876510 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPINB9P1 | NR_033851.1 | |||
| BC025340.1 | AAH25340.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830575 | Chr.6:2847376 - 3035682 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76458597] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map