Assay Details
Target Gene Details
Entrez Gene ID: | 80350 |
Gene Name: | lipoprotein(a) like 2, pseudogene |
Gene Aliases: |
APOA2, APOAL, APOARGC, apo(a)rg-C |
Location: |
Chr.6:160466555-160511124 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LPAL2 | NR_028092.1 | |||
| NR_028093.1 | ||||
| U19517.1 | AAA85692.1 | |||
| U19518.1 | AAA85693.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3611438 | Chr.6:160487993 - 160503876 on Build GRCh38 | Gain |
|
| nsv5566 | Chr.6:160484923 - 160518346 on Build GRCh38 | Insertion |
|
| esv35098 | Chr.6:160001557 - 160649838 on Build GRCh38 | Gain |
|
| nsv818463 | Chr.6:160014537 - 160604515 on Build GRCh38 | Gain |
|
| esv6519 | Chr.6:160456601 - 160535561 on Build GRCh38 | Loss |
|
| nsv970186 | Chr.6:160497203 - 160511769 on Build GRCh38 | Duplication |
|
| esv3611436 | Chr.6:160456712 - 160535559 on Build GRCh38 | Gain |
|
| esv2733055 | Chr.6:160457077 - 160668796 on Build GRCh38 | Deletion |
|
| nsv1026543 | Chr.6:160306836 - 161585804 on Build GRCh38 | Gain |
|
| esv2759484 | Chr.6:159951212 - 160813633 on Build GRCh38 | Gain+Loss |
|
| nsv525990 | Chr.6:160482590 - 160549931 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116058816,rs116513039] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map