Assay Details
Target Gene Details
Entrez Gene ID: | 728464 |
Gene Name: | methyltransferase like 24 |
Gene Aliases: |
C6orf186, dJ71D21.2 |
Location: |
Chr.6:110245946-110358272 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| METTL24 | NM_001123364.1 | NP_001116836.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2671329 | Chr.6:110248533 - 110250682 on Build GRCh38 | Deletion |
|
| esv25776 | Chr.6:110248521 - 110250688 on Build GRCh38 | Loss |
|
| esv3571359 | Chr.6:110248341 - 110252617 on Build GRCh38 | Loss |
|
| esv3540306 | Chr.6:110248508 - 110250712 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map