Assay Details
Target Gene Details
Entrez Gene ID: | 3663 |
Gene Name: | interferon regulatory factor 5 |
Gene Aliases: |
SLEB10 |
Location: |
Chr.7:128937737-128950042 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2670312 | Chr.7:128499519 - 128999613 on Build GRCh38 | Deletion |
|
| esv2735137 | Chr.7:128499440 - 128999664 on Build GRCh38 | Deletion |
|
| nsv608415 | Chr.7:128867243 - 128954129 on Build GRCh38 | Loss |
|
| nsv831125 | Chr.7:128772651 - 128955697 on Build GRCh38 | Loss |
|
| esv22837 | Chr.7:128774435 - 128953151 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map