Assay Details
Target Gene Details
Entrez Gene ID: | 6561 |
Gene Name: | solute carrier family 13 member 1 |
Gene Aliases: |
NAS1, NaSi-1 |
Location: |
Chr.7:123113490-123201836 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC13A1 | NM_001324400.1 | NP_001311329.1 | ||
| NM_022444.3 | NP_071889.2 | |||
| XM_011516515.2 | XP_011514817.1 | |||
| XM_011516516.2 | XP_011514818.1 | |||
| XM_011516517.2 | XP_011514819.1 | |||
| XM_017012554.1 | XP_016868043.1 | |||
| AF260824.1 | AAG60583.1 | |||
| AK026413.1 | BAB15477.1 | |||
| AK314537.1 | ||||
| BC111775.1 | AAI11776.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6605n100 | Chr.7:122924680 - 123134549 on Build GRCh38 | Gain |
|
| nsv1019131 | Chr.7:123056169 - 123181026 on Build GRCh38 | Gain |
|
| nsv1033520 | Chr.7:122726490 - 123352883 on Build GRCh38 | Loss |
|
| nsv524192 | Chr.7:122763367 - 123578468 on Build GRCh38 | Gain |
|
| esv3891198 | Chr.7:121885430 - 123886905 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map