Assay Details
Target Gene Details
Entrez Gene ID: | 613126 |
Gene Name: | CYP51A1 antisense RNA 1 |
Gene Aliases: |
LRRD1-AS1 |
Location: |
Chr.7:92134563-92180725 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP51A1-AS1 | NR_122109.1 | |||
| NR_122110.1 | ||||
| AL133568.1 | ||||
| HY053824.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3614137 | Chr.7:92170742 - 92180166 on Build GRCh38 | Loss |
|
| esv2734818 | Chr.7:92170750 - 92180166 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs138400949] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map