Assay Details
Target Gene Details
Entrez Gene ID: | 78996 |
Gene Name: | chromosome 7 open reading frame 49 |
Gene Aliases: |
MRI |
Location: |
Chr.7:135092303-135170826 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C7orf49 | NM_001305630.1 | NP_001292559.1 | ||
| XM_017012595.1 | XP_016868084.1 |
Target Gene Details
Entrez Gene ID: | 55281 |
Gene Name: | transmembrane protein 140 |
Gene Aliases: |
- |
Location: |
Chr.7:135148014-135166215 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM140 | NM_018295.4 | NP_060765.4 | ||
| AK001862.1 | BAA91948.1 | |||
| AK225055.1 | ||||
| AK301268.1 | ||||
| BC020942.1 | AAH20942.1 | |||
| DB064371.1 | ||||
| DC323016.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527174 | Chr.7:134952791 - 135157183 on Build GRCh38 | Loss |
|
| nsv527014 | Chr.7:135149118 - 135206907 on Build GRCh38 | Gain |
|
| esv3615115 | Chr.7:135148990 - 135156413 on Build GRCh38 | Loss |
|
| esv2665250 | Chr.7:135149068 - 135156387 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73725242] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map