Assay Details
Target Gene Details
Entrez Gene ID: | 3614 |
Gene Name: | inosine monophosphate dehydrogenase 1 |
Gene Aliases: |
IMPD, IMPD1, IMPDH-I, LCA11, RP10, sWSS2608 |
Location: |
Chr.7:128392277-128409988 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv949940 | Chr.7:128378147 - 128405346 on Build GRCh38 | Deletion |
|
| nsv608400 | Chr.7:128338766 - 128408140 on Build GRCh38 | Loss |
|
| nsv831124 | Chr.7:128330292 - 128507856 on Build GRCh38 | Gain+Loss |
|
| esv2493565 | Chr.7:128345641 - 128413323 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map