Assay Details
Target Gene Details
Entrez Gene ID: | 100128081 |
Gene Name: | JAZF1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.7:28180457-28241377 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JAZF1-AS1 | NR_034097.1 | |||
| AK293540.1 | ||||
| DB481568.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830931 | Chr.7:28132064 - 28329752 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73091254,rs76228795] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map