Assay Details
Target Gene Details
Entrez Gene ID: | 11333 |
Gene Name: | PDGFA associated protein 1 |
Gene Aliases: |
HASPP28, PAP, PAP1 |
Location: |
Chr.7:99394673-99408682 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDAP1 | NM_014891.6 | NP_055706.1 | ||
| BC000684.2 | AAH00684.1 | |||
| BC007873.2 | AAH07873.1 | |||
| BG281467.1 | ||||
| BI460344.1 | ||||
| U41745.1 | AAC50462.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831069 | Chr.7:99350505 - 99529961 on Build GRCh38 | Loss |
|
| nsv831068 | Chr.7:99221949 - 99408858 on Build GRCh38 | Loss |
|
| nsv951379 | Chr.7:99375678 - 99405377 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map