Assay Details
Target Gene Details
Entrez Gene ID: | 55665 |
Gene Name: | upregulator of cell proliferation |
Gene Aliases: |
URG4 |
Location: |
Chr.7:43875894-43926411 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| URGCP | NM_001077664.2 | NP_001071132.1 | ||
| NM_001290076.1 | NP_001277005.1 | |||
| AK302459.1 | ||||
| BG722670.1 | ||||
| CD628752.1 | ||||
| DC336174.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428165 | Chr.7:43855828 - 44172960 on Build GRCh38 | Gain |
|
| nsv830980 | Chr.7:43865998 - 44078539 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map