Assay Details
Target Gene Details
Entrez Gene ID: | 8887 |
Gene Name: | Tax1 binding protein 1 |
Gene Aliases: |
CALCOCO3, T6BP, TXBP151 |
Location: |
Chr.7:27739373-27829767 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAX1BP1 | NM_001079864.2 | NP_001073333.1 | ||
| NM_001206901.1 | NP_001193830.1 | |||
| NM_001206902.1 | NP_001193831.1 | |||
| NM_006024.6 | NP_006015.4 | |||
| XM_005249900.2 | XP_005249957.1 | |||
| XM_017012754.1 | XP_016868243.1 | |||
| AF090891.1 | AAF24016.1 | |||
| AF268075.1 | AAG03025.1 | |||
| AK000098.1 | ||||
| AK296720.1 | ||||
| AK299729.1 | ||||
| AK301064.1 | ||||
| AK314292.1 | ||||
| AL136586.1 | CAB66521.1 | |||
| BC050358.2 | AAH50358.1 | |||
| BP280714.1 | ||||
| CR457056.1 | CAG33337.1 | |||
| CR533556.1 | CAG38587.1 | |||
| DB455556.1 | ||||
| DC424592.1 | ||||
| U33821.2 | AAA75595.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759520 | Chr.7:27753997 - 27933250 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map