Assay Details
Target Gene Details
Entrez Gene ID: | 2645 |
Gene Name: | glucokinase |
Gene Aliases: |
FGQTL3, GK, GLK, HHF3, HK4, HKIV, HXKP, LGLK, MODY2 |
Location: |
Chr.7:44143213-44189439 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GCK | NM_000162.3 | NP_000153.1 | ||
| NM_033507.1 | NP_277042.1 | |||
| NM_033508.1 | NP_277043.1 | |||
| XM_017011966.1 | XP_016867455.1 | |||
| AK122876.1 | ||||
| BC001890.1 | AAH01890.1 | |||
| BT007062.1 | AAP35711.1 | |||
| DA640823.1 | ||||
| EU008605.1 | ||||
| M69051.1 | AAB59563.1 | |||
| M88011.1 | AAA51824.1 | |||
| M90299.1 | AAA52562.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830983 | Chr.7:44068842 - 44201624 on Build GRCh38 | Loss |
|
| nsv8082 | Chr.7:43956156 - 44207229 on Build GRCh38 | Gain |
|
| nsv428165 | Chr.7:43855828 - 44172960 on Build GRCh38 | Gain |
|
| nsv509195 | Chr.7:44147118 - 44261782 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map