Assay Details
Target Gene Details
Entrez Gene ID: | 9603 |
Gene Name: | nuclear factor, erythroid 2 like 3 |
Gene Aliases: |
NRF3 |
Location: |
Chr.7:26152227-26187137 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFE2L3 | NM_004289.6 | NP_004280.5 | ||
| AB010812.1 | BAA76288.1 | |||
| AF133059.1 | AAG43275.1 | |||
| AF134891.1 | AAF61404.1 | |||
| BC015336.1 | AAH15336.1 | |||
| BC049219.1 | AAH49219.1 | |||
| BC056142.1 | AAH56142.1 | |||
| BC068455.1 | AAH68455.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891090 | Chr.7:26011300 - 26194682 on Build GRCh38 | Gain |
|
| esv3612615 | Chr.7:25958361 - 26200719 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map