Assay Details
Target Gene Details
Entrez Gene ID: | 340351 |
Gene Name: | ATP/GTP binding protein like 3 |
Gene Aliases: |
CCP3 |
Location: |
Chr.7:134985169-135135782 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGBL3 | NM_178563.3 | NP_848658.3 | ||
| XM_017012136.1 | XP_016867625.1 | |||
| XM_017012137.1 | XP_016867626.1 | |||
| XM_017012141.1 | XP_016867630.1 | |||
| AK023045.1 | BAB14371.1 | |||
| AK302827.1 | ||||
| AL832281.1 | 1 | 1184 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527174 | Chr.7:134952791 - 135157183 on Build GRCh38 | Loss |
|
| esv3891208 | Chr.7:134986279 - 134993772 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map