Assay Details
Target Gene Details
Entrez Gene ID: | 54476 |
Gene Name: | ring finger protein 216 |
Gene Aliases: |
CAHH, TRIAD3, U7I1, UBCE7IP1, ZIN |
Location: |
Chr.7:5620041-5781730 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF216 | NM_207111.3 | NP_996994.1 | ||
| NM_207116.2 | NP_996999.1 | |||
| XM_005249785.2 | XP_005249842.1 | |||
| XM_011515436.1 | XP_011513738.1 | |||
| XM_017012363.1 | XP_016867852.1 | |||
| XM_017012364.1 | XP_016867853.1 | |||
| XM_017012365.1 | XP_016867854.1 | |||
| AF228527.1 | AAF36723.1 | |||
| AF513717.1 | AAP47174.1 | |||
| AF513718.1 | AAP47175.1 | |||
| AK000916.1 | BAA91422.1 | |||
| AK092767.1 | ||||
| AK123635.1 | ||||
| AK292396.1 | ||||
| AK297231.1 | ||||
| AK297416.1 | ||||
| AK314332.1 | ||||
| AY062174.1 | AAL38043.1 | |||
| AY177396.1 | AAO60361.1 | |||
| AY177397.1 | AAO60362.1 | |||
| AY177398.1 | AAO60363.1 | |||
| BC000787.2 | AAH00787.2 | |||
| BC063825.1 | AAH63825.1 | |||
| BX537406.1 | CAD97648.1 |
Target Gene Details
Entrez Gene ID: | 100874342 |
Gene Name: | RNF216 intronic transcript 1 |
Gene Aliases: |
- |
Location: |
Chr.7:5662432-5680461 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF216-IT1 | NR_046834.1 | |||
| AA825874.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv436904 | Chr.7:5035574 - 6713282 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map