Assay Details
Target Gene Details
Entrez Gene ID: | 392843 |
Gene Name: | IQ motif containing with AAA domain 1 like |
Gene Aliases: |
IQCA1P1 |
Location: |
Chr.7:151190873-151205784 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IQCA1L | NM_001304419.1 | NP_001291348.1 | ||
| HM370405.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831188 | Chr.7:151038117 - 151215789 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map