Assay Details
Target Gene Details
Entrez Gene ID: | 10447 |
Gene Name: | family with sequence similarity 3 member C |
Gene Aliases: |
GS3786, ILEI |
Location: |
Chr.7:121348851-121396369 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM3C | NM_001040020.1 | NP_001035109.1 | ||
| NM_014888.2 | NP_055703.1 | |||
| XM_011515736.2 | XP_011514038.1 | |||
| XM_011515737.2 | XP_011514039.1 | |||
| AK290682.1 | ||||
| AK292731.1 | ||||
| BC024200.1 | AAH24200.1 | |||
| BC046932.1 | AAH46932.1 | |||
| BC068526.1 | AAH68526.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759558 | Chr.7:121268405 - 121353272 on Build GRCh38 | Loss |
|
| nsv1034353 | Chr.7:121178328 - 121400522 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map