Assay Details
Target Gene Details
Entrez Gene ID: | 4191 |
Gene Name: | malate dehydrogenase 2 |
Gene Aliases: |
M-MDH, MDH, MGC:3559, MOR1 |
Location: |
Chr.7:76048019-76067508 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MDH2 | NM_001282403.1 | NP_001269332.1 | ||
| NM_001282404.1 | NP_001269333.1 | |||
| NM_005918.3 | NP_005909.2 | |||
| NR_104165.1 | 2 | 449 | ||
| AF047470.1 | AAC03787.1 | |||
| AI793009.1 | 2 | 344 | ||
| AK095803.1 | ||||
| AK290779.1 | ||||
| AK293460.1 | ||||
| AK316587.1 | ||||
| BC001917.1 | AAH01917.1 | |||
| BM463639.1 | 2 | 299 | ||
| BP382246.1 | ||||
| CR536548.1 | CAG38785.1 | |||
| DC354622.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428173 | Chr.7:75934531 - 76173362 on Build GRCh38 | Gain |
|
| nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain |
|
| esv3613754 | Chr.7:76045674 - 76060185 on Build GRCh38 | Gain |
|
| dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map