Assay Details
Target Gene Details
Entrez Gene ID: | 155006 |
Gene Name: | transmembrane protein 213 |
Gene Aliases: |
- |
Location: |
Chr.7:138797487-138806759 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM213 | NM_001085429.1 | 3 | 2903 | NP_001078898.1 |
| XM_005250167.3 | 3 | 3407 | XP_005250224.1 | |
| AI672404.1 | ||||
| AW771283.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831154 | Chr.7:138785743 - 138989367 on Build GRCh38 | Gain |
|
| esv2735212 | Chr.7:138060002 - 140415947 on Build GRCh38 | Deletion |
|
| esv3615199 | Chr.7:138793218 - 138823621 on Build GRCh38 | Gain |
|
| esv3615200 | Chr.7:138794105 - 138836379 on Build GRCh38 | Gain |
|
| nsv1030956 | Chr.7:138478424 - 138894773 on Build GRCh38 | Gain |
|
| nsv516706 | Chr.7:138801791 - 138809843 on Build GRCh38 | Loss |
|

Genomic Map