Assay Details
Target Gene Details
Entrez Gene ID: | 10552 |
Gene Name: | actin related protein 2/3 complex subunit 1A |
Gene Aliases: |
Arc40, HEL-68, HEL-S-307, SOP2Hs, SOP2L |
Location: |
Chr.7:99325873-99366262 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARPC1A | NM_001190996.1 | NP_001177925.1 | ||
| NM_006409.3 | NP_006400.2 | |||
| AB209059.1 | BAD92296.1 | |||
| AK223015.1 | BAD96735.1 | |||
| BC039594.1 | AAH39594.2 | |||
| BC047889.1 | AAH47889.2 | |||
| BC054027.1 | AAH54027.1 | |||
| EU668326.1 | ||||
| GQ472234.1 | ||||
| Y08999.1 | CAA70203.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831069 | Chr.7:99350505 - 99529961 on Build GRCh38 | Loss |
|
| nsv5863 | Chr.7:99349505 - 99373187 on Build GRCh38 | Insertion |
|
| nsv831068 | Chr.7:99221949 - 99408858 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map