Assay Details
Target Gene Details
Entrez Gene ID: | 80228 |
Gene Name: | ORAI calcium release-activated calcium modulator 2 |
Gene Aliases: |
C7orf19, CBCIP2, MEM142B, TMEM142B |
Location: |
Chr.7:102433530-102456821 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ORAI2 | NM_001126340.2 | NP_001119812.1 | ||
| NM_001271818.1 | NP_001258747.1 | |||
| NM_001271819.1 | NP_001258748.1 | |||
| NM_032831.3 | NP_116220.1 | |||
| AK027639.1 | BAB55256.1 | |||
| AK056497.1 | ||||
| AK300574.1 | ||||
| BC069270.1 | AAH69270.1 | |||
| CN366874.1 | ||||
| DA770788.1 | ||||
| DC384892.1 | ||||
| HY064081.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831083 | Chr.7:102340560 - 102655422 on Build GRCh38 | Loss |
|
| nsv428182 | Chr.7:102257460 - 102735503 on Build GRCh38 | Gain |
|
| dgv6538n100 | Chr.7:102171110 - 103063688 on Build GRCh38 | Gain |
|
| nsv607998 | Chr.7:102385213 - 102473462 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map