Assay Details
Target Gene Details
Entrez Gene ID: | 613126 |
Gene Name: | CYP51A1 antisense RNA 1 |
Gene Aliases: |
LRRD1-AS1 |
Location: |
Chr.7:92134563-92180725 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP51A1-AS1 | NR_122109.1 | 5 | 2268 | |
| NR_122110.1 | 3 | 1750 | ||
| AL133568.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3614137 | Chr.7:92170742 - 92180166 on Build GRCh38 | Loss |
|
| esv2734818 | Chr.7:92170750 - 92180166 on Build GRCh38 | Deletion |
|

Genomic Map