Assay Details
Target Gene Details
Entrez Gene ID: | 221938 |
Gene Name: | monocyte to macrophage differentiation associated 2 |
Gene Aliases: |
PAQR10 |
Location: |
Chr.7:4892245-4959213 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MMD2 | NM_001100600.1 | NP_001094070.1 | ||
| NM_001270375.1 | NP_001257304.1 | |||
| NM_198403.3 | NP_940685.3 | |||
| NR_072989.1 | ||||
| AK094524.1 | ||||
| AY424288.1 | AAR08376.1 | |||
| BC037881.1 | AAH37881.2 | |||
| BC067905.1 | AAH67905.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv605980 | Chr.7:4836426 - 4963742 on Build GRCh38 | Loss |
|
| nsv1161486 | Chr.7:4907246 - 4927827 on Build GRCh38 | Duplication |
|
| nsv1019120 | Chr.7:4890022 - 5098778 on Build GRCh38 | Loss |
|
| dgv6238n100 | Chr.7:4810816 - 5030133 on Build GRCh38 | Loss |
|
| esv2763613 | Chr.7:4914210 - 4955400 on Build GRCh38 | Loss |
|
| nsv528748 | Chr.7:4780950 - 4971362 on Build GRCh38 | Loss |
|
| nsv8033 | Chr.7:4762656 - 5046966 on Build GRCh38 | Loss |
|
| dgv1249e214 | Chr.7:4861563 - 4973875 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map