Assay Details
Target Gene Details
Entrez Gene ID: | 346562 |
Gene Name: | G protein subunit alpha transducin 3 |
Gene Aliases: |
GDCA |
Location: |
Chr.7:80457291-80512503 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNAT3 | NM_001102386.2 | NP_001095856.1 | ||
| XM_011516124.2 | XP_011514426.1 | |||
| BC147016.1 | ||||
| BC147017.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831042 | Chr.7:80354763 - 80492996 on Build GRCh38 | Gain |
|
| esv3613860 | Chr.7:80453738 - 80584904 on Build GRCh38 | Loss |
|
| dgv1305e214 | Chr.7:80401473 - 80470340 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map