Assay Details
Target Gene Details
Entrez Gene ID: | 54467 |
Gene Name: | ankyrin repeat and IBR domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.7:92246216-92401773 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKIB1 | NM_019004.1 | NP_061877.1 | ||
| XM_005250458.3 | XP_005250515.1 | |||
| XM_011516334.1 | XP_011514636.1 | |||
| XM_017012359.1 | XP_016867848.1 | |||
| AB037807.1 | ||||
| BC063861.1 | AAH63861.1 |
More Information
Set Membership: |
|

Genomic Map