Assay Details
Target Gene Details
Entrez Gene ID: | 653857 |
Gene Name: | ARP3 actin-related protein 3 homolog C |
Gene Aliases: |
ARP11 |
Location: |
Chr.7:149881359-150325523 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACTR3C | NM_001164458.1 | NP_001157930.1 | ||
| XM_005250043.4 | XP_005250100.3 | |||
| XM_011516502.2 | XP_011514804.1 | |||
| XM_011516506.2 | XP_011514808.1 | |||
| XM_011516507.2 | XP_011514809.1 | |||
| XM_011516508.2 | XP_011514810.1 | |||
| XM_011516511.2 | XP_011514813.1 | |||
| XM_017012548.1 | XP_016868037.1 | |||
| XM_017012549.1 | XP_016868038.1 | |||
| XM_017012550.1 | XP_016868039.1 | |||
| XM_017012551.1 | XP_016868040.1 | |||
| XM_017012552.1 | XP_016868041.1 | |||
| XM_017012553.1 | XP_016868042.1 | |||
| BX640643.1 | CAI56711.1 | |||
| DA868104.1 | ||||
| DN992136.1 | ||||
| DR004860.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1034409 | Chr.7:150147550 - 150749983 on Build GRCh38 | Gain |
|
| nsv1015582 | Chr.7:150270897 - 150588016 on Build GRCh38 | Gain |
|
| nsv508490 | Chr.7:150308979 - 150354152 on Build GRCh38 | Deletion |
|
| nsv608990 | Chr.7:150264185 - 151010400 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map