Assay Details
Target Gene Details
Entrez Gene ID: | 64764 |
Gene Name: | cAMP responsive element binding protein 3 like 2 |
Gene Aliases: |
BBF2H7 |
Location: |
Chr.7:137874979-138002101 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CREB3L2 | NM_001318246.1 | NP_001305175.1 | ||
| NM_194071.3 | NP_919047.2 | |||
| XM_005250541.4 | XP_005250598.1 | |||
| XM_017012540.1 | XP_016868029.1 | |||
| XM_017012541.1 | XP_016868030.1 | |||
| AJ549092.1 | CAD79342.1 | |||
| AJ549387.1 | CAD79347.1 | |||
| AK131517.1 | 10 | 1957 | BAD18659.1 | |
| AK293048.1 | ||||
| BC110813.1 | AAI10814.1 | |||
| BC131723.1 | ||||
| BX649143.1 | ||||
| CR749563.1 | CAH18360.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5963 | Chr.7:137877442 - 137911305 on Build GRCh38 | Insertion |
|
| nsv1022793 | Chr.7:137502637 - 137934613 on Build GRCh38 | Gain |
|
| nsv831150 | Chr.7:137815813 - 137968062 on Build GRCh38 | Gain+Loss |
|
| esv33947 | Chr.7:137235266 - 138283104 on Build GRCh38 | Gain |
|
| nsv518070 | Chr.7:137849881 - 138124093 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map