Assay Details
Target Gene Details
Entrez Gene ID: | 80727 |
Gene Name: | tweety family member 3 |
Gene Aliases: |
- |
Location: |
Chr.7:2631969-2664802 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTYH3 | NM_025250.2 | NP_079526.1 | ||
| XM_011515535.2 | XP_011513837.1 | |||
| XM_017012655.1 | XP_016868144.1 | |||
| XM_017012656.1 | XP_016868145.1 | |||
| AB051478.2 | ||||
| AB162931.1 | BAD20190.1 | |||
| AL522452.3 | ||||
| BC131824.1 | ||||
| BC152447.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv464257 | Chr.7:2629953 - 2705490 on Build GRCh38 | Loss |
|
| nsv950793 | Chr.7:2592167 - 2703366 on Build GRCh38 | Deletion |
|
| nsv605863 | Chr.7:2612996 - 2655983 on Build GRCh38 | Loss |
|
| nsv5618 | Chr.7:2626956 - 2657598 on Build GRCh38 | Insertion |
|
| nsv471227 | Chr.7:2589386 - 2729230 on Build GRCh38 | Loss |
|
| dgv11145n54 | Chr.7:2628411 - 2659000 on Build GRCh38 | Loss |
|
| nsv1161479 | Chr.7:2614158 - 2673300 on Build GRCh38 | Deletion |
|
| nsv1027694 | Chr.7:2585778 - 2669607 on Build GRCh38 | Gain |
|
| dgv11144n54 | Chr.7:2620317 - 2712853 on Build GRCh38 | Loss |
|
| nsv471238 | Chr.7:2623184 - 2669580 on Build GRCh38 | Gain |
|
| dgv401n21 | Chr.7:2600507 - 2714033 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map