Assay Details
Target Gene Details
Entrez Gene ID: | 8326 |
Gene Name: | frizzled class receptor 9 |
Gene Aliases: |
CD349, FZD3 |
Location: |
Chr.7:73433779-73436120 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FZD9 | NM_003508.2 | 1 | 898 | NP_003499.1 |
| BC026333.1 | 1 | 894 | AAH26333.1 | |
| U82169.1 | 1 | 694 | AAC51174.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759537 | Chr.7:72446485 - 73461568 on Build GRCh38 | Gain+Loss |
|
| nsv831027 | Chr.7:73173922 - 73442066 on Build GRCh38 | Loss |
|
| dgv6467n100 | Chr.7:73420075 - 73583448 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map